Automatic burettes, Schilling pattern, PMMA, class B
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7.010.125 7.010.150
661-2405EA 105 EUR
661-2405 661-2410
Automatic burettes, Schilling pattern, PMMA, class B
Buretten
Automatic burettes, class DIN-B, short line graduation, clear plastic (PMMA).
  • Push button enabling a precise titration
  • Complete with polyethylene reservoir
  • Included with a stable base
  • Automatic zero point
Liquid delivery time 30 to 60 seconds, without waiting time.