Dishes, evaporating, quartz glass
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8.651.001 8.651.004 8.651.006 8.651.007 8.651.005
WITG8.651.001EA 0 EUR
WITG8.651.001 WITG8.651.004 WITG8.651.006 WITG8.651.007 WITG8.651.005
Dishes, evaporating, quartz glass
Reservoirs/Schalen/Potten Uitdampschalen
Evaporating dishes, made of quartz glass, with spout.
  • Excellent chemical resistance
  • Excellent temperature resistance (up to 1000 °C)
Certificaten: DIN 12341