Laboratorium-/mediaflessen, smalle hals, rond, borosilicaatglas, met schroefdop
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41330112 41330118 41330124 41330130 41330148 41330142 41330136
HECH41330112EA 5.5 EUR
HECH41330112 HECH41330118 HECH41330124 HECH41330130 HECH41330148 HECH41330142 HECH41330136
Laboratorium-/mediaflessen, smalle hals, rond, borosilicaatglas, met schroefdop
Flessen Glazen flessen
Borosilicate glass 3,3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Round, with graduations, DIN GL 45 thread, pouring ring and blue PP screw cap.
Order Now
Specificaties
Environmentally Preferable