Laboratory bottles, wide neck, with screw cap and GL 45 thread
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41331130 41331124
HECH41331130EA 44.3 EUR
HECH41331130 HECH41331124
Laboratory bottles, wide neck, with screw cap and GL 45 thread
Flessen Glazen flessen
Borosilicate glass, amber, coated.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With screw cap and pouring ring.