Assortments of tube end fittings, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F704-04 F704-02
BOHLF704-04EA 125 EUR
BOHLF704-04 BOHLF704-02
Assortments of tube end fittings, BOLA
Bevestigingen Schroeven
PP, coloured.
  • Temperature resistance: from 0 to +100 °C
  • Very good chemical resistance
  • Different colours for better distinction
  • FDA conform
Tube end fittings made of PP in 9 different colours, two fittings per colour. With male thread UNF ¼" 28 G; washers made of PA are included in delivery.Colours: natural, black, orange, yellow, green, blue, violet and grey.
Order Now
Specificaties
Environmentally Preferable