Connectors, tubing, miniature couplings
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F707-08 F707-04 F707-16 F707-02 F707-06 F707-20 F707-14 F707-18
BOHLF707-08EA 11.7 EUR
BOHLF707-08 BOHLF707-04 BOHLF707-16 BOHLF707-02 BOHLF707-06 BOHLF707-20 BOHLF707-14 BOHLF707-18
Connectors, tubing, miniature couplings
Fittingen en connectoren
Miniature tubing connectors, made of fluoroplastic, with female thread for making branchings.