3-Isobutyl-1-methylxanthine
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
13347-100 13347-250 13347-500
CAYM13347-100EA 57.1 EUR
CAYM13347-100 CAYM13347-250 CAYM13347-500
3-Isobutyl-1-methylxanthine
3-Isobutyl-1-methylxanthine
Formule: C₁₀H₁₄N₄O₂
Smeltpunt: 200…201 °C
MDL Nummer: MFCD00005584
CAS nummer: 28822-58-4
EINECS: 249-259-3