2-Aminobenzamidoxime

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-Amino-N-hydroxybenzimidamide

11062-500 11062-1 11062-5
CAYM11062-500EA 54.9 EUR
CAYM11062-500 CAYM11062-1 CAYM11062-5
2-Aminobenzamidoxime
2-Aminobenzamidoxime
Formule: C₇H₉N₃O
MDL Nummer: MFCD00492723
CAS nummer: 16348-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR