Diethyl-tert-butylmalonate ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
L09131.06
L09131.14
L09131.06EA
133
EUR
L09131.06
L09131.14
Diethyl-tert-butylmalonate ≥97%
Diethyl-tert-butylmalonate
Purity: 97%
CAS No.: 759-24-0
Molecular Formula: C11H20O4
Formula Weight: 216.28
Flash Point: 90°(194°F)
Refractive Index: 1.4250
Boiling Point: 102-104°/11mm
Beilstein Reference: 1781709
MDL No.: MFCD00009152
Formule:
C₁₁H₂₀O₄ MW: 216.28 g/mol Kookpunt: 102…104 °C (11 mmHg) Dichtheid: 0.978 Vlampunt: 90 °C (194 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009152 CAS nummer: 759-24-0 EINECS: 212-067-5 |