Ethyl-4-tert-butylbenzoylformate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SPB07813.250 SPB07813.1
MAYBSPB07813.250EA 0 EUR
MAYBSPB07813.250 MAYBSPB07813.1
Ethyl-4-tert-butylbenzoylformate
Ethyl-4-tert-butylbenzoylformate
Formule: C₁₄H₁₈O₃
CAS nummer: 80120-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR