Bis(pentamethylcyclopentadienyl)iron(II)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Bis(pentamethylcyclopentadienyl)iron, Decamethylferrocene
347740010 347740050
ACRO347740010EA 123 EUR
ACRO347740010 ACRO347740050
Bis(pentamethylcyclopentadienyl)iron(II)
Bis(pentamethylcyclopentadienyl)iron(II)
Formule: C₂₀H₃₀Fe
MW: 326.30 g/mol
Smeltpunt: 277 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00058736
CAS nummer: 12126-50-0
Order Now
Specification Test Results
Appearance Yellow to orange Crystalline powder
NMR (1H-NMR) Consistent with structure