4-Bromophenyl octyl ether ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A13334.14
A13334.03
A13334.14EA
372
EUR
A13334.14
A13334.03
4-Bromophenyl octyl ether ≥98%
4-Bromophenyl octyl ether
Purity: 98%
CAS No.: 96693-05-9
Molecular Formula: C14H21BrO
Formula Weight: 285.23
Refractive Index: 1.5180
Boiling Point: 135-137°/0.5mm
MDL No.: MFCD00043445
Formule:
C₁₄H₂₁BrO Kookpunt: 135…137 °C (0,5 mmHg) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00043445 CAS nummer: 96693-05-9 |