Diethyl mesoxalate (diethyl ketomalonate) 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Diethyl 2-oxomalonate
A12771.06 A12771.14 A12771.22
A12771.06EA 45.7 EUR
A12771.06 A12771.14 A12771.22
Diethyl mesoxalate (diethyl ketomalonate) 95%
Diethyl mesoxalate (diethyl ketomalonate)
Purity: 95% CAS No.: 609-09-6 Molecular Formula: C7H10O5 Formula Weight: 174.15 Flash Point: 110°(230°F) Refractive Index: 1.4150 Melting Point: ca -70° Boiling Point: 208-210° Beilstein Reference: 971873 Merck Reference: 14,5913 MDL No.: MFCD00009121
Formule: C₇H₁₀O₅
MW: 174.15 g/mol
Kookpunt: 208…210 °C
Smeltpunt: ∼–70 °C
Dichtheid: 1.142
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009121
CAS nummer: 609-09-6
EINECS: 210-176-2