Di-tert-butyl malonate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
406620100 406620500
ACRO406620100EA 49.2 EUR
ACRO406620100 ACRO406620500
Di-tert-butyl malonate 98%
Di-tert-butyl malonate
Formule: C₁₁H₂₀O₄
Kookpunt: 110…111 °C (22 mmHg)
Smeltpunt: –7 °C
Dichtheid: 0,966 g/cm³
Vlampunt: 88 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008810
CAS nummer: 541-16-2
EINECS: 208-769-6
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4175 to 1.4195 (20°C, 589 nm)
GC ≥97.5 %