(1,2,2-Trimethylpropylidene)malononitrile
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KM03717.1
KM03717.50
MAYBKM03717.1EA
0
EUR
MAYBKM03717.1
MAYBKM03717.50
(1,2,2-Trimethylpropylidene)malononitrile
(1,2,2-Trimethylpropylidene)malononitrile
Formule:
C₉H₁₂N₂ |
CAS nummer:
13017-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...