O-Isobutylhydroxylamine hydrochloride ≥98.0% (by total nitrogen basis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
I0387-1G
I0387-5G
TCIAI0387-1GEA
129
EUR
TCIAI0387-1G
TCIAI0387-5G
O-Isobutylhydroxylamine hydrochloride ≥98.0% (by total nitrogen basis)
O-Isobutylhydroxylamine hydrochloride
Formule:
C₄H₁₂ClNO Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00060208 CAS nummer: 6084-58-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...