Tris(trimethylsilyl)phosphite ≥96%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Tris(trimethylsilyl)phosphite
17159.02
17159.04
17159.02EA
30.1
EUR
17159.02
17159.04
Tris(trimethylsilyl)phosphite ≥96%
Tris(trimethylsilyl)phosphite
Purity: 96%
CAS No.: 1795-31-9
Molecular Formula: [(CH3)3SiO]3P
Formula Weight: 298.54
Physical Form: Liquid
Density: 0.893
Refractive Index: 1.409
Sensitivity: Air and Moisture Sensitive
Boiling Point: 80°/8mm
MDL No.: MFCD00015588
Formule:
C₉H₂₇O₃PSi₃ MW: 298.54 g/mol Kookpunt: 80 °C (8 mmHg) Dichtheid: 0.893 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00015588 CAS nummer: 1795-31-9 |