(10S,17S)-DiHDoHE
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10008128-25 10008128-50 10008128-100
CAYM10008128-25EA 277 EUR
CAYM10008128-25 CAYM10008128-50 CAYM10008128-100
(10S,17S)-DiHDoHE
(10S,17S)-DiHDoHE
Formule: C₂₂H₃₂O₄
CAS nummer: 871826-47-0