2-(1-Piperidinyl)aniline
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
2-(1-Piperidino)aniline
BTB02217DA
BTB02217EA
BTB02217.50
MAYBBTB02217DAEA
0
EUR
MAYBBTB02217DA
MAYBBTB02217EA
MAYBBTB02217.50
2-(1-Piperidinyl)aniline
2-(1-Piperidinyl)aniline
Formule:
C₁₁H₁₆N₂ MW: 176.26 g/mol |
MDL Nummer:
MFCD00047467 CAS nummer: 39643-31-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...