2-Amino-5-phenylpyrazine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A1684-1G A1684-5G
TCIAA1684-1GEA 71.5 EUR
TCIAA1684-1G TCIAA1684-5G
2-Amino-5-phenylpyrazine ≥98.0% (by GC, titration analysis)
2-Amino-5-phenylpyrazine
Formule: C₁₀H₉N₃
MDL Nummer: MFCD01646266
CAS nummer: 13535-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4b56afb7e3ce0d636043c0dc057c3d2fe59fbb5&vid=c4b56afb7e3ce0d636043c0dc057c3d2fe59fbb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR