4-(1H-1,2,4-Triazol-1-yl)phenol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4-(1,2,4-Triazol-1-yl)phenol

AC18635.1 AC18635EA AC18635EE
MAYBAC18635.1EA 0 EUR
MAYBAC18635.1 MAYBAC18635EA MAYBAC18635EE
4-(1H-1,2,4-Triazol-1-yl)phenol
4-(1H-1,2,4-Triazol-1-yl)phenol
Formule: C₈H₇N₃O
MW: 161.16 g/mol
MDL Nummer: MFCD00003096
CAS nummer: 68337-15-5
EINECS: 269-845-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR