Drum funnels
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DRM672-BK DRM452-BK
NEWPDRM672-BKEA 148 EUR
NEWPDRM672-BK NEWPDRM452-BK
Drum funnels
Trechters Filtertrechters Trechters voor vaten
Thick, double-wall polyethylene construction resists UV rays, corrosion and most chemicals.
  • Allows access to ³/₄" bung for use of liquid-level gauge
  • Level inside surface allows easy draining of containers, parts or filters
  • Zigzag ridges prevent splashes and catch parts or tools
These large diameter funnels covers entire drum top to minimize drips and spills.