Drum fill gauge
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DRM245 DRM317
NEWPDRM245EA 29 EUR
NEWPDRM245 NEWPDRM317
Drum fill gauge
Niveausensoren
These durable gauge helps prevent overfilling by popping up when drum is near capacity.
  • Corrosion-resistant polyethylene
  • Highly visible red and yellow offers easy indication
  • Avoid overflows and downtime by quickly checking a drum's liquid level