Eppendorf® Xplorer® Plus elektronische meerkanaalspipetten
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4861000.767 4861000.775 4861000.783 4861000.791 4861000.805 4861000.813 4861000.821 4861000.830 4861000.778 4861000.779 4861000.792 4861000.793
613-2235EA 1250 EUR
613-2235 613-2236 613-2237 613-2238 613-2239 613-2240 613-2241 613-2242 613-6986 613-6987 613-6990 613-6991
Eppendorf® Xplorer® Plus elektronische meerkanaalspipetten
Pipetten Micropipetten
Eppendorf Xplorer plus multi-channel pipettes offer the ideal solution for precise, efficient liquid handling in high-throughput applications, with ergonomic design, a user-friendly interface, and programmable modes for quick, reliable pipetting, enhancing reproducibility and minimising errors.
  • Multifunctional rocker: 'Up is up and down is down™' principle
  • Clearly arranged color display; no sub-menus; help function
  • Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
  • ON/OFF switch: Switches the pipetter off during longer periods of inactivity
  • Programming: One-time programming of sequences for easier handling of complex processes
  • Individual fixed volume setting: Desired volume can be saved
  • Password protection: Protects programming and adjustment
  • Individual speed setting: Sets parameters to your individual working speed
Reliably reproducible results:
Tips always rest in the same position
Volume and speed precisely and reproducibly adjustable
Adjustable to your work environment and requirements
PhysioCare Concept™:
Reduced weight
Improved ergonomics
Perfect balance
Optimally angled pipetter head
Improved view and high resolution display
Spring-loaded tip cone provides maximum tightness
Pipetter maintenance reminder function, based on time or frequency of use as desired. Operate in the language of your choice. And has an easy selection of all functions with the practical selection dial.
Air-cushion pipetting system, with autoclavable lower part.
Applications: Pipetting of aqueous liquids, pipetting liquids (manual and automatic), dispensing (timed and manual) and pipetting and mixing. Additional applications include aspirating, diluting, sequential dispensing and reverse pipetting.