Capillary columns, Elite-CLPesticides2 for autosystem
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9316668 N9316670
PERCN9316668EA 1150 EUR
PERCN9316668 PERCN9316670
Capillary columns, Elite-CLPesticides2 for autosystem
Kolommen voor chromatografie
Elite-CL Pesticides 2 capillary columns are specially designed to overcome the coelutions and analyte breakdown typically encountered in chlorinated pesticide.