Capillary columns, Elite-Alumina for autosystem
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9316544 N9316304 N9316305
PERCN9316544EA 3140 EUR
PERCN9316544 PERCN9316304 PERCN9316305
Capillary columns, Elite-Alumina for autosystem
Kolommen voor chromatografie
Elite-Alumina PLOT phase capillary columns are ideal for the analysis of light hydrocarbons.