Adapting centring rings for small flange connections
Leverancier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20660127 20660128 20660129
185-1281EA 24.3 EUR
185-1281 185-1282 185-1283
Adapting centring rings for small flange connections
Afdichtingen Afdichtringen
These adapting centring rings for small flange connections with sealing ring made of FKM.