Microwave digestion vessels, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A240-02 A240-06 A240-08 A240-10 A244-06 A244-04 A244-02
BOHLA240-02EA 527 EUR
BOHLA240-02 BOHLA240-06 BOHLA240-08 BOHLA240-10 BOHLA244-06 BOHLA244-04 BOHLA244-02
Microwave digestion vessels, BOLA
Destructiesystemen Mineralisatiewerkstations
Dimensionally stable basic vessel and screw cover made of TFM, homogenous, non porous surface.
  • One piece of sealing and rupture membranes already mounted
  • For samples of up to maximum 0,5 g
The liner allows a more precise weighted sample and different digestions with only one basic vessel.
Leveringsinformatie: Includes 10 pieces of replacement membranes.