Elbow pieces with small flanges KF
Leverancier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20669400 20669404 20669405 20669406 20673400 20673414 20673415 20673416
185-2401EA 45.8 EUR
185-2401 185-2402 185-2403 185-2404 185-2411 185-2412 185-2413 185-2414
Elbow pieces with small flanges KF
Fittingen en connectoren
These elbow pieces with small flanges are made of aluminium or stainless steel.
Used for vacuum system.