Craft handles and blades, Professional range
Leverancier: SWANN MORTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9220 9210
SWAN9220EA 266 EUR
SWAN9220 SWAN9210
Craft handles and blades, Professional range
Scalpels Scalpelhandvaten
These handles and blades are ideal for artwork, craft work and modelling.
  • Surgical blades are made from quality steel
  • Cutting edge performance meets the demands of professionals
  • Blades protected in anti-corrosion packaging