Blood collection tubes with EDTA anticoagulant, vacuum, Vacutest®
Leverancier: KIMA SAS DI CHIARIN RENZO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13030 13125 135400 13560 13040 13001 13005 13505 13060 135100 135300 13010 13510
720-2742EA 251 EUR
720-2742 KIMA13125 720-2745 720-2770 720-2743 720-2765 720-2766 720-2768 720-2767 KIMA135100 KIMA135300 KIMA13010 KIMA13510
Blood collection tubes with EDTA anticoagulant, vacuum, Vacutest®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The test tubes are made from Polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use, size 40×20 mm, printed in three colours.
  • Available in different sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes