Test tubes, with screw cap
Leverancier: KIMA SAS DI CHIARIN RENZO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17122 17174
KIMA17122EA 579 EUR
KIMA17122 KIMA17174
Test tubes, with screw cap
Buisjes Reageerbuisjes
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
Universal tubes for a wide range of applications.