Test tubes, with screw cap
Leverancier: KIMA SAS DI CHIARIN RENZO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17040 17042 17048 17050 17056 17058 17022 17004 17018
212-6711EA 457 EUR
212-6711 212-6712 212-6715 212-6716 212-6717 212-6718 KIMA17022 KIMA17004 KIMA17018
Test tubes, with screw cap
Buisjes Reageerbuisjes
PP, translucent or PS, clear, with screw cap.
  • PS: Good thermal and chemical resistance
  • PP: Excellent thermal and chemical resistance, autoclavable
  • Robust, economical
Universal tubes for a wide range of applications.