Test tubes with rim
Leverancier: KIMA SAS DI CHIARIN RENZO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18016 18304 18020 18008
212-6721EA 0 EUR
212-6721 212-6726 212-0183 KIMA18008
Test tubes with rim
Buisjes Reageerbuisjes
PP, translucent or PS, clear, with rim.
  • PS: Good thermal and chemical resistance
  • PP: Excellent thermal and chemical resistance, autoclavable
  • Robust, economical
Universal tubes for a wide range of applications.