Blind small flanges
Leverancier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20669000 20669004 20669005 20669006 20671000 20671004 20671005 20671006
185-2101EA 12.7 EUR
185-2101 185-2102 185-2103 185-2104 185-2111 185-2112 185-2113 185-2114
Blind small flanges
Fittingen en connectoren
Blind small flanges KF made of aluminium or stainless steel.
Used for vacuum system.