Robotic tips for Eppendorf epMotion® automated pipetting system
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
965-261 965-021 965-262 966-021 966-261 966-262 967-021 967-261 967-262
736-0755EA 0 EUR
736-0755 736-0756 736-0757 736-0758 736-0759 736-0760 736-0761 736-0762 736-0763
Robotic tips for Eppendorf epMotion® automated pipetting system
Pipettips
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
  • Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
  • Tips have a low percentage Coefficient of Variation (% CV)
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen