Funnels with short stem
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
49050003 49050006 49050009 49050012 49050015 49050018 49050021
HECH49050003EA 1.5 EUR
HECH49050003 HECH49050006 HECH49050009 HECH49050012 221-0316 HECH49050018 HECH49050021
Funnels with short stem
Trechters Filtertrechters Filtertrechters
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
  • Good chemical resistance
  • Suspension eye for easy storage
  • Easy to clean