Centrifuge tubes, cylindrical, short conical bottom
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40948002 40948001
HECH40948002EA 79.5 EUR
HECH40948002 HECH40948001
Centrifuge tubes, cylindrical, short conical bottom
Buisjes Centrifugebuisjes
Borosilicate glass 3.3.
  • Ungraduated or graduated
  • Excellent chemical resistance
  • Excellent temperature resistance
Short conical bottom.