Centrifuge tubes, cylindrical, round bottom
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40950001 40950003 40950016 40950002 40951002 40951003 40951001
525-1312EA 38.5 EUR
525-1312 525-1316 525-1322 HECH40950002 HECH40951002 HECH40951003 HECH40951001
Centrifuge tubes, cylindrical, round bottom
Buisjes Centrifugebuisjes
AR-Glas® (soda-lime glass).
  • Ungraduated or graduated
  • Good chemical resistance
Round bottom, rimless.