Centrifuge tubes, cylindrical, round bottom
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40958016 40959004 40959002 40959003 40959001 40958004
HECH40958016EA 59.8 EUR
HECH40958016 HECH40959004 HECH40959002 HECH40959003 HECH40959001 HECH40958004
Centrifuge tubes, cylindrical, round bottom
Buisjes Centrifugebuisjes
Borosilicate glass 3.3.
  • Ungraduated or graduated
  • Excellent chemical resistance
  • Excellent temperature resistance
Round bottom, rimless