Powder funnels
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44002044 44002030
HECH44002044EA 26.2 EUR
HECH44002044 HECH44002030
Powder funnels
Trechters Poedertrechters
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable