Bottle pourers
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
028.030 028.032 028.038 028.045BLUE
COWI028.030EA 60.8 EUR
COWI028.030 COWI028.032 COWI028.038 COWI028.045BLUE
Bottle pourers
Trechters Druppeltrechters
Manufactured in pure, FDA approved, PTFE.
  • Totally chemically resistant and inert
  • To fit GL45, 38, 32 and 30 screw neck bottles
  • State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.