Bellow connectors
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008.1414 008.1919 008.2424 008.2929 008.1919.1
COWI008.1414EA 44.8 EUR
COWI008.1414 COWI008.1919 COWI008.2424 COWI008.2929 COWI008.1919.1
Bellow connectors
Adapters Adapters met NS-verbindingen
Bellow connector, PTFE, for use as a flexible or elastic connector to prevent pressure and stresses, and to dampen vibrations in standard ground joints.
  • Inert
  • Temperature resistant up to 280 °C
  • Excellent chemical resistance
With ground joints on cone and socket. Sturdy construction with trapezoidal sealing rings, which give medium to high vacuum performance.