Syringes for HPLC autosamplers, for Spectra-Physics
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
202145 202192
HAMI202145EA 127 EUR
HAMI202145 HAMI202192
Syringes for HPLC autosamplers, for Spectra-Physics
Spuiten Injectiespuiten voor chromatografie
These gas tight syringes are designed for HPLC autosamplers.
  • Temperature range: 20 to 50 °C
  • Thread size 1/4-28