Separating funnels, Squibb
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42986060 42986048 42985024 42985030 42985048 42985060 42985066 42985072
HECH42986060EA 68.7 EUR
HECH42986060 HECH42986048 HECH42985024 HECH42985030 HECH42985048 HECH42985060 HECH42985066 HECH42985072
Separating funnels, Squibb
Trechters Scheitrechters
Separating funnels, Squibb, made of borosilicate glass 3.3, with solid glass plug or plug with PTFE key and plastic stopper.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance