Stoppers for standard ground joints, octogonal, HDPE
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42990007 42990010 42990012 42990014 42990019 42990024 42990029 42990034 42990045
HECH42990007EA 0.6 EUR
HECH42990007 HECH42990010 HECH42990012 HECH42990014 HECH42990019 HECH42990024 HECH42990029 HECH42990034 HECH42990045
Stoppers for standard ground joints, octogonal, HDPE
Stoppen PE/LDPE/HDPE stoppen
High-density polyethylene.
  • Acid and alkali resistant
  • Octogonal