Watchmakers' magnifiers
Leverancier: ESCHENBACH OPTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1130 11245 11247 1124110
ESCH1130EA 49.9 EUR
ESCH1130 ESCH11245 ESCH11247 ESCH1124110
Watchmakers' magnifiers
Vergrootglazen en loepen
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
  • Includes vent to prevent condensation
  • Made of flame resistant plastic material
  • Colour: anthracite