Miniature luer connectors, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F717-02 F717-06 F717-04 F717-08
BOHLF717-02EA 28.9 EUR
BOHLF717-02 BOHLF717-06 BOHLF717-04 BOHLF717-08
Miniature luer connectors, BOLA
Fittingen en connectoren
Connector made of PTFE for transition from Luer connection to female thread UNF ¼" 28 G.
  • Temperature resistance from -200 °C to +250 °C
  • Universal chemical resistance