Transition fittings, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F760-08 F760-18 F760-04 F760-14
BOHLF760-08EA 65.6 EUR
BOHLF760-08 BOHLF760-18 BOHLF760-04 BOHLF760-14
Transition fittings, BOLA
Fittingen en connectoren
Connectors made of PTFE, one side with metric thread and connecting nut with compression rings for connecting tubing or tubes with O.D. 4, 6, 8 or 10 mm. Other side with female thread UNF ¼" 28 G for connecting flanged tubing with O.D. 1,6 or 3,2 mm.
  • Temperature resistance from –20 to +120 ºC
  • Excellent chemical resistance
  • FDA conformity