Deactivated/silinised vials
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5182-0567 9301-0977 5188-2788 5188-5390
HEWL5182-0567EA 29 EUR
HEWL5182-0567 HEWL9301-0977 HEWL5188-2788 HEWL5188-5390
Deactivated/silinised vials
flesjes
These vials made of PP are used for pH sensitive samples, sodium or heavy metals analysis.
  • Translucent
  • Good chemical resistance
  • Can be used with crimp or snap caps
Vials with glass insert are used for economical and effective microsampling. The PP body has a glass flanged insert molded to the inside, so the sample comes in contact only with the 100 µl glass insert and the septum.
Order Now
Specificaties
Environmentally Preferable